missing translation for 'onlineSavingsMsg'
Learn More

Melford Bicine

Product Code. p-200086995
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 30018117

Brand: Melford B74060100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zwitterionic buffer used in a variety of molecular biology applications such as protein resolution, protein crystallization and SDS-PAGE.

Zwitterionic buffer used in a variety of molecular biology applications such as protein resolution, protein crystallization and SDS-PAGE.

pKa 25°C: 8.3

pH Range: 7.6-9.0

Melting Range: 190-192°C

Specifications

Chemical Name or Material Bicine
CAS 150-25-4
Quantity 100 g
Molecular Formula C6H13NO4
Grade Ultra Pure
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.