missing translation for 'onlineSavingsMsg'
Learn More

Bicine, 99.38%, MP Biomedicals™

Product Code. 11318589 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
Product Code. Quantity unitSize
11318589 25 g 25g
11328589 100 g 100g
11338589 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 11318589

Brand: MP Biomedicals 0210100525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-bis [2-hydroxyethyl]glycine
PubChem CID 8761
ChEBI CHEBI:40957
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 0.9938
IUPAC Name sodium 2-[bis(2-hydroxyethyl)amino]acetate
Formula Weight 163.2
Chemical Name or Material Bicine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.