missing translation for 'onlineSavingsMsg'
Learn More

Bicine, 99+%, for analysis

Product Code. 10742564 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
This item is not returnable. View return policy

Product Code. 10742564

Brand: Thermo Scientific Acros 172652500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Absorbance 0.05 max. (5% aqueous soln.) at 250nm
Absorbance Conditions (5% aq. soln.) (1cm cell)
CAS Min % 99.0
CAS Max % 100.0
Color White
pH 4.0 to 5.5 (10% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula (HOCH2CH2)2NCH2CO2H
Packaging Glass bottle
Merck Index 15, 1205
Formula Weight 163.17
Percent Purity ≥99.0% (Total base)
Grade Analytical
Chemical Name or Material Bicine, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.