missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bicine, 99+%, for biochemistry

Product Code. 10129512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10129512 25 g Plastic bottle 25g
10153441 100 g Plastic bottle 100g
10331104 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10129512 Supplier Thermo Scientific Chemicals Supplier No. 327710250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Absorbance 0.05 max. (0.5M, water) at 260nm, 0.05 max. (0.5M, water) at 280nm
Melting Point 190.0°C to 192.0°C
CAS Min % 99.0
CAS Max % 100.0
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula (HOCH2CH2)2NCH2CO2H
Packaging Plastic bottle
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in ethanol, ether, chloroform and, benzene
Formula Weight 163.17
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material Bicine, For biochemistry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.