missing translation for 'onlineSavingsMsg'
Learn More

Bicine, Fisher BioReagents

Product Code. 10638203
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
250g
This item is not returnable. View return policy

Product Code. 10638203

Brand: Fisher BioReagents BP2646100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Bicine is one of the zwitterionic amino acids known as Good buffers. Used as a biological buffer and chelating agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-bis [2-hydroxyethyl]glycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Absorbance 0.100 AU max. at 260nm, 0.100 AU max. at 280nm
Melting Point 190°C
Color White
pH 4.0 to 5.0
Physical Form Powder or Solid
Quantity 100 g
Linear Formula (HOCH2CH2)2NCH2COOH
Packaging Poly Bottle
Merck Index 15, 1205
Formula Weight 163.2
Chemical Name or Material Bicine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.