missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Bicine is used in a thin layer ion exchange chromatography method for protein resolution
- It is used in peptide and protein crystallization
- Bicine is used in a kinetic study of a quaternary transition-state analogue complex of creatine kinase and in multiphasic buffer system for SDS-PAGE of proteins
Specifications
Specifications
| CAS | 150-25-4 |
| Melting Point | 190°C (literature) |
| Color | White |
| Physical Form | Powder |
| Quantity | 1 kg |
| Molecular Formula | C6H12NNaO4 |
| Linear Formula | C6H13NO4 |
| MDL Number | MFCD00004295 |
| Synonym | bicine, n,n-bis 2-hydroxyethyl glycine, diethylolglycine, dihydroxyethylglycine, diethanol glycine, n,n-di 2-hydroxyethyl glycine, bicene, 2-bis 2-hydroxyethyl amino acetic acid, n,n-dihydroxyethyl glycine, n,n-dihydroxyethylglycine |
| Solubility Information | Soluble in water (10mg/mL-clear, colorless solution; a saturated solution in water is 1.1 M ) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?