missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (-)-Bicuculline methiodide

Product Code. 30564780
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30564780 50 mg 50mg
18702554 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30564780 Supplier Tocris Bioscience™ Supplier No. 2503/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

GABAA antagonist; more water soluble version of (+)-bicuculline (Cat. No. 0130)

Methiodide form of classical GABAA receptor antagonist (+)-bicuculline . More water-soluble and stable. Non-GABA receptor-mediated actions reported, including actions on calcium-dependent potassium channels.

Methobromide Salt and Methochloride Salt also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40709-69-1
Molecular Formula C21H20INO6

Specifications

Quantity 50 mg
Formula Weight 509.3
Purity >99%
Molecular Weight (g/mol) 509.296
Chemical Name or Material (-)-Bicuculline methiodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.