missing translation for 'onlineSavingsMsg'
Learn More

Bicyclohexyl, 98%

Product Code. 11445563
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11445563

Brand: Thermo Scientific Alfa Aesar A13481.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-51-3 | C12H22 | 166.31 g/mol

Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-51-3
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00003815
InChI Key WVIIMZNLDWSIRH-UHFFFAOYSA-N
Synonym bicyclohexyl, 1,1'-bicyclohexyl, dicyclohexyl, bicyclohexane, dicyclohexane, dodecahydrobiphenyl, cyclohexane, cyclohexyl, 1,1'-biphenyl, dodecahydro, 1,1'-bi cyclohexyl, 1, dodecahydro
PubChem CID 7094
IUPAC Name cyclohexylcyclohexane
SMILES C1CCC(CC1)C1CCCCC1

Spezifikation

Melting Point 3°C to 4°C
Density 0.864
Boiling Point 226°C to 228°C
Flash Point 92°C (198°F)
Refractive Index 1.478
Quantity 10 g
Beilstein 1848266
Solubility Information Difficult to mix.
Formula Weight 166.31
Percent Purity 98%
Chemical Name or Material Bicyclohexyl
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt