missing translation for 'onlineSavingsMsg'
Learn More

Bifenazate, TRC

Product Code. 30008992
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008992 25 g 25g
30006438 10 g 10g
17982575 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008992 Supplier Toronto Research Chemicals Supplier No. TRCB38302025G
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149877-41-8
Molecular Formula C17 H20 N2 O3
Molecular Weight (g/mol) 300.35
Synonym 1-Methylethyl 2-(4-methoxy[1,1'-biphenyl]-3-yl)-hydrazinecarboxylate, Acramite, Bifenazate, D 2341, Floramite, Isopropyl 3-(4-methoxy-3-biphenylyl)carbazate
IUPAC Name propan-2-yl N-(2-methoxy-5-phenylanilino)carbamate
SMILES COc1ccc(cc1NNC(=O)OC(C)C)c2ccccc2

Specifications

Analyte or Component Names Bifenazate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C17H20N2O3/c1-12(2)22-17(20)19-18-15-11-14(9-10-16(15)21-3)13-7-5-4-6-8-13/h4-12,18H,1-3H3,(H,19,20)
Solution Type Neat
Industry Type Environmental Testing
Quantity 25 g
Formula Weight 300.1474
Percent Purity >95%
Chemical Name or Material Bifenazate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.