missing translation for 'onlineSavingsMsg'
Learn More

Bifonazole, 98%, Thermo Scientific™

Product Code. 15530584 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15530584 25 g 25g
15510584 1 g 1g
15520584 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15530584 Supplier Thermo Scientific Acros Supplier No. 458620250
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 60628-96-8
Molecular Formula C22H18N2
Molecular Weight (g/mol) 310.39
InChI Key OCAPBUJLXMYKEJ-UHFFFAOYSA-N
Synonym bifonazole, mycospor, trifonazole, bifonazol, bifonazolum, amycor, azolmen, bifonazol inn-spanish, bifonazolum inn-latin, 1-4-biphenylyl phenylmethyl-1h-imidazole
PubChem CID 2378
ChEBI CHEBI:78692
IUPAC Name 1-[phenyl-(4-phenylphenyl)methyl]imidazole
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4

Specifications

Melting Point 149.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 1 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 310.39
Percent Purity 98%
Chemical Name or Material Bifonazole
compliance-icons
Product Identifier
  • Bifonazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.