missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bikunin Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149397
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
| Bikunin | |
| Polyclonal | |
| Unconjugated | |
| A1M; Alpha-1 microglycoprotein; Alpha-1-microglobulin; alpha-1-microglobulin/bikunin precursor; AMBP; Bikunin; complex-forming glycoprotein heterogeneous in charge; EDC1; growth-inhibiting protein 19; HCP; HI30; HI-30; IATIL; Inter-alpha-trypsin inhibitor light chain; ITI; ITIL; ITILC; ITI-LC; protein AMBP; Protein HC; trypstatin; uristatin; uronic-acid-rich protein; UTI | |
| Mouse myeloma cell line NS0-derived recombinant human Bikunin, Ala206-Asn352. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction