Learn More
Invitrogen™ BIN1 Recombinant Rabbit Monoclonal Antibody (23GB3010), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553343
 View more versions of this product
Product Code. 30089234
  • 207.00 € / 20µL
Description
This gene encodes several isoforms of a nucleocytoplasmic adaptor protein, one of which was initially identified as a MYC-interacting protein with features of a tumor suppressor. Isoforms that are expressed in the central nervous system may be involved in synaptic vesicle endocytosis and may interact with dynamin, synaptojanin, endophilin, and clathrin. Isoforms that are expressed in muscle and ubiquitously expressed isoforms localize to the cytoplasm and nucleus and activate a caspase-independent apoptotic process. Studies in mouse suggest that this gene plays an important role in cardiac muscle development. Alternate splicing of the gene results in ten transcript variants encoding different isoforms. Aberrant splice variants expressed in tumor cell lines have also been described.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BIN1
Recombinant Monoclonal
Unconjugated
BIN1
ALP-1; AMPH 2; Amph2; amphiphysin 2; amphiphysin II; amphiphysin IIamph2; amphiphysin-like; amphiphysin-like protein; AMPHL; Bin1; box dependant MYC interacting protein 1; box-dependent myc-interacting protein 1; BRAMP-2; bridging integrator 1; DKFZp547F068; MGC10367; MGC105358; myc box dependent interacting protein 1; myc box-dependent-interacting protein 1; SH3 domain-containing protein 9; SH3P9; similar to bridging integrator 1 isoform 9
Rabbit
Affinity chromatography
RUO
117028, 274, 30948
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB3010
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O00499, O08539, O08839
BIN1
A synthesized peptide derived from human BIN1 (400-593AA).
20 μL
Primary
Human, Mouse, Rat