missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 572.00 €
Specifications
| Antigen | BIN2 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639098
|
Novus Biologicals
NBP2-48690-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627888
|
Novus Biologicals
NBP2-48690 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BIN2 Polyclonal antibody specifically detects BIN2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| BIN2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.2), 40% Glycerol | |
| 51411 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BRAP1, BRAP-1, breast cancer associated protein BRAP1, Breast cancer-associated protein 1, bridging integrator 2, bridging integrator-2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EKPVRTPEAKENENIHNQNPEELCTSPTLMTSQVASEPGEAKKMEDKEKDNKLISADSSEGQDQLQVSMVPENNNLTAPEPQEEVSTSENPQL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title