missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ 4 HPLC Columns

Product Code. 10581105 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4 mm
4.6 mm
Unit Size:
Each
Pack of 4
This item is not returnable. View return policy

Product Code. 10581105

Brand: Thermo Scientific™ 72305102130

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use this 300Å wide pore silica for analysis of larger hydrophobic proteins and peptides that bind too tightly on C8 or C18 columns.

Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Length (Metric) 100 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 9
Pore Size 300 Å
Surface Area 100 m²/g
Temperature 60°C
USP Type L26
Stationary Phase C4
Product Line BioBasic
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 4%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type Analytical Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.