missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ 4 HPLC Columns

Product Code. 10583995 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4 mm
4.6 mm
Unit Size:
Each
Pack of 4
Product Code. Length (Metric) Diameter (Metric) unitSize
10583995 50 mm 1 mm Each
10263313 50 mm 2.1 mm Each
10395003 100 mm 1 mm Each
10581105 100 mm 2.1 mm Each
10692854 100 mm 4.6 mm Each
10649585 150 mm 1 mm Each
10110693 150 mm 2.1 mm Each
10384513 150 mm 4.6 mm Each
10344563 250 mm 2.1 mm Each
10541305 250 mm 4.6 mm Each
11598320 10 mm 2.1 mm Pack of 4
11508330 10 mm 4 mm Pack of 4
12 options
This item is not returnable. View return policy

Product Code. 10583995

Brand: Thermo Scientific™ 72305051030

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use this 300Å wide pore silica for analysis of larger hydrophobic proteins and peptides that bind too tightly on C8 or C18 columns.

Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 1 mm
Length (Metric) 50 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 9
Pore Size 300 Å
Surface Area 100 m²/g
Temperature 60°C
USP Type L26
Stationary Phase C4
Product Line BioBasic
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 4%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type Analytical Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.