missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ AX HPLC Columns

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

Brand:  Thermo Scientific™ 73105-151030

971.30 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.



Code : 50

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10170703

  • 1022.42 € / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

  • 5μm particle size, 300Å pore size
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
  • Low hydrophobicity
  • Capacity of 0.22mEq/g; 3% carbon load
  • Use for normal phase and hydrophilic interaction chromatography (HILIC)
  • Stable under demanding pH conditions (2 to 8 pH range)
Specifications

Specifications

Analytical Column
Low
5800 psi (400 bar)
2 to 8
100 m²/g
N/A
BioBasic
3%
No
Bio-LC
1 mm
150 mm
5 μm
300 Å
60°C
Polyethyleneimine
Spherical, Fully Porous, Ultrapure Silica
Ion Exchange
Ion Exchange
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.