missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ AX HPLC Column

Product Code. 16140781 Shop All Thermo Scientific Products
Change view
Click to view available options
Particle Size:
5 μm
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size Unit Size
16140781 5 μm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16140781 Supplier Thermo Scientific™ Supplier No. 73105022151
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.

  • 5 μm particle size, 300 Å pore size
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine bound to pure deactivated silica
  • Low hydrophobicity
  • Capacity of 0.22 mEq/g; 3% carbon load
  • Suitable for normal phase and hydrophilic interaction chromatography (HILIC)
  • Stable under demanding pH conditions (2 to 8 pH range)
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 5 μm
Length (Metric) 20 mm
Type Guard Catridge
Diameter (Metric) Inner 2.1 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.