missing translation for 'onlineSavingsMsg'
Learn More

Biochanin A, TRC

Product Code. 30135315
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135315 1 g 1g
30137779 250 mg 250mg
17955065 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135315 Supplier Toronto Research Chemicals Supplier No. TRCB3872501G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-80-5
Molecular Formula C16 H12 O5
Molecular Weight (g/mol) 284.26
Synonym 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-one (ACI), Biochanin A (6CI), Isoflavone, 5,7-dihydroxy-4'-methoxy- (8CI), (-)-5,7-Dihydroxy-4'-methoxyisoflavone, 4'-Methylgenistein, 5,7-Dihydroxy-4'-methoxyisoflavone, Biochanin, Genistein 4-methyl ether, Genistein 4'-methyl ether, NSC 123538, Olmelin
IUPAC Name 5,7-dihydroxy-3-(4-methoxyphenyl)chromen-4-one
SMILES COc1ccc(cc1)C2=COc3cc(O)cc(O)c3C2=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H12O5/c1-20-11-4-2-9(3-5-11)12-8-21-14-7-10(17)6-13(18)15(14)16(12)19/h2-8,17-18H,1H3
Formula Weight 284.0685
Percent Purity >95%
Chemical Name or Material Biochanin A
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.