missing translation for 'onlineSavingsMsg'
Learn More

Benchmark Scientific™ BioClave Autoclave Printer

Product Code. 17954432
Change view
Click to view available options
For Use With (Equipment):
BioClave Research Autoclaves
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17954432 BioClave Research Autoclaves Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17954432 Supplier Benchmark Scientific™ Supplier No. B4000P
  • ONLINE EXCLUSIVE PRICE
    397.00 €
    341.12 € / Pack of 1
    Save 55.88 €
    14% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The BioClave Autoclave printer can be attached to the autoclave to provide a printout of runs for documentation.

  • Connects to all BioClave Research Autoclaves
  • Provides documentation of autoclave process

Specifications

For Use With (Application) Printing Out Run Parameters
For Use With (Equipment) BioClave Research Autoclaves
Model BioClave
Weight (English) 1 lb.
Weight (Metric) 2.2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.