missing translation for 'onlineSavingsMsg'
Learn More

Biocytin, MedChemExpress

Product Code. 16492796
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492796 25 mg 25mg
16483628 10 mg 10mg
16434716 100 mg 100mg
16410027 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492796 Supplier Medchem Express Supplier No. HY101884
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Biocytin is a conjugate of D-biotin and L-lysine, where the carboxylate of D-biotin is coupled with the ε-amine of L-lysine via a secondary amide bond. Biocytin is a classical neuroanatomical tracer commonly used to map brain connectivity. Biocytin is used as a versatile marker in anterograde, retrograde and intracellular neuroanatomical investigations and in biotinidase assays.

Biocytin is a conjugate of D-biotin and L-lysine, where the carboxylate of D-biotin is coupled with the ε-amine of L-lysine via a secondary amide bond. Biocytin is a classical neuroanatomical tracer commonly used to map brain connectivity. Biocytin is used as a versatile marker in anterograde, retrograde and intracellular neuroanatomical investigations and in biotinidase assays.

Specifications

Chemical Name or Material Biocytin
Physical Form Solid
CAS 576-19-2
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C16H28N4O4S
Synonym (+)-Biocytin
Solubility Information H2O : 50 mg/mL (134.24 mM; Need ultrasonic)
SMILES N[C@@H](CCCCNC(CCCC[C@@H]1SC[C@]([C@]1([H])N2)([H])NC2=O)=O)C(O)=O
Molecular Weight (g/mol) 372.48
Formula Weight 372.48
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.