missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biodiesel Blank, VHG™
Description
Biodiesel Blank
Specifications
Specifications
| Chemical Name or Material | Biodiesel Blank |
| Methods | ICP |
| Analyte or Component Names | Biodiesel Blank |
| Shipping Condition | Room Temperature |
| IUPAC Name | Biodiesel Blank |
| Solvent or Matrix | B100 Biodiesel |
| Quantity | 100 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?