missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin (Vitamin B7 or Vitamin H) Antibody (SPM375), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34771JF646
This item is not returnable.
View return policy
Description
Biotin (Vitamin B7 or Vitamin H) Monoclonal antibody specifically detects Biotin (Vitamin B7 or Vitamin H) in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Biotin (Vitamin B7 or Vitamin H) | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H | |
| Biotinylated sheep immunoglobulin | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPM375 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction