missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-carboxylic acid, 98%

Product Code. 11459653
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459653 25 g 25g
11469653 100 g 100g
11479653 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459653 Supplier Thermo Scientific Alfa Aesar Supplier No. A12438.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 92-92-2 | C13H10O2 | 198.221 g/mol

Biphenyl-4-carboxylic acid is employed in the synthesis, characterization of europium, terbium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4-carboxylic acid is employed in the synthesis, characterization of europium, terbium complexes.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-92-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00002553
InChI Key NNJMFJSKMRYHSR-UHFFFAOYSA-N
Synonym 4-biphenylcarboxylic acid, biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, p-phenylbenzoic acid, 4-carboxybiphenyl, para phenyl benzoic acid, 4-diphenylcarboxylic acid, diphenyl-4-carboxylic acid, 4-carboxy-1,1'-biphenyl
PubChem CID 66724
IUPAC Name 4-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 223°C to 227°C
Odor Odorless
Quantity 25 g
Beilstein 973519
Solubility Information Insoluble in water.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material Biphenyl-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.