missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Birch pollen Bet V1 Antibody (M0418) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-09013-0.2mg
This item is not returnable.
View return policy
Description
Birch pollen Bet V1 Monoclonal antibody specifically detects Birch pollen Bet V1 in Plant samples. It is validated for ELISA, X-Ray Crystallography
Specifications
| Birch pollen Bet V1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Plant | |
| Purified |
| ELISA, X-Ray Crystallography | |
| M0418 | |
| Unconjugated | |
| Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A | |
| Bet v 1-A, a major allergen of the European white birch. | |
| 0.2 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction