missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-aminophenyl) ether, 98%

Product Code. 15461618
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461618 1 g 1g
15481618 250 mg 250mg
15471618 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15461618 Supplier Thermo Scientific Alfa Aesar Supplier No. H55396.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 24878-25-9 | C12H12N2O | 200.241 g/mol

It is catalytic reagent and used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is catalytic reagent and used in pharmaceutical industry.

Solubility
Slightly soluble in water.

Notes
Store at low temperature, away from light and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24878-25-9
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD00792533
InChI Key GOJFAKBEASOYNM-UHFFFAOYSA-N
Synonym 2,2'-oxydianiline, 2-2-aminophenoxy aniline, 2,2 inverted exclamation marka-oxydianiline, aminophenyl ether, o,o'-oxydianiline, 2-aminophenyl ether, bis 2-aminophenyl ether, 2,2'-oxybis benzenamine, benzenamine,2,2'-oxybis, 2,2'-diaminodiphenyl ether
PubChem CID 458824
IUPAC Name 2-(2-aminophenoxy)aniline
SMILES C1=CC=C(C(=C1)N)OC2=CC=CC=C2N

Specifications

Melting Point 61°C to 64°C
Color Brown to White
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 200.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bis(2-aminophenyl) ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.