missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl)ether-4,4'-dihydroxydiphenyl Sulfone Copolymer, TRC

Product Code. 30011476
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011476 1 g 1g
30006986 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011476 Supplier Toronto Research Chemicals Supplier No. TRCB4189701G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190078-71-8
Molecular Formula (C16 H16 O5 S)n C12 H10 O4 S
Synonym alpha-[4-[(4-hydroxyphenyl)sulfonyl]phenyl]-omega-hydroxy-Poly(oxy-1,2-ethanediyloxy-1,2-ethanediyloxy-1,4-phenylenesulfonyl-1,4-phenylene)
IUPAC Name 4,4'-(((oxybis(ethane-2,1-diyl))bis(oxy))bis(4,1-phenylenesulfonyl))diphenol polymer
SMILES O=S(C1=CC=C(O)C=C1)(C2=CC=C(OCCOCCOC3=CC=C(S(C4=CC=C(O)C=C4)(=O)=O)C=C3)C=C2)=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C28H26O9S2/c29-21-1-9-25(10-2-21)38(31,32)27-13-5-23(6-14-27)36-19-17-35-18-20-37-24-7-15-28(16-8-24)39(33,34)26-11-3-22(30)4-12-26/h1-16,29-30H,17-20H2
Chemical Name or Material Bis(2-chloroethyl)ether-4,4'-dihydroxydiphenyl Sulfone Copolymer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.