missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) terephthalate, 97%

Product Code. p-67996 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 10732202

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 6422-86-2 | C24H38O4 | 390.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6422-86-2
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.55
MDL Number MFCD00072256
InChI Key RWPICVVBGZBXNA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl terephthalate, kodaflex dotp, di-2-ethylhexyl terephthalate, 1,4-benzenedicarboxylic acid, bis 2-ethylhexyl ester, terephthalic acid, bis 2-ethylhexyl ester, eastman 168, ccris 7052, di 2-ethylhexyl terephthalate, bis 2-ethylhexyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-bis 2-ethylhexyl ester
PubChem CID 22932
IUPAC Name bis(2-ethylhexyl) benzene-1,4-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)C(=O)OCC(CC)CCCC

Specifications

CAS Min % 96
CAS Max % 100.0
Melting Point -67.0°C
Color Colorless
Density 0.9840g/mL
Boiling Point 400.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4890 to 1.4910
Quantity 250 g
Beilstein 09, IV, 3306
Specific Gravity 0.984
Solubility Information Solubility in water: insoluble
Formula Weight 390.55
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) terephthalate, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.