missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) terephthalate, 97%, Thermo Scientific™

Product Code. 15537039 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 kg
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15537039 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15537039 Supplier Thermo Scientific Acros Supplier No. 402490100
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6422-86-2
Melting Point -67.0°C
Density 0.9840g/mL
Boiling Point 400.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C24H38O4
Refractive Index 1.4890 to 1.4910
Quantity 10 kg
Beilstein 09,IV,3306
Specific Gravity 0.984
Synonym bis 2-ethylhexyl terephthalate, kodaflex dotp, di-2-ethylhexyl terephthalate, 1,4-benzenedicarboxylic acid, bis 2-ethylhexyl ester, terephthalic acid, bis 2-ethylhexyl ester, eastman 168, ccris 7052, di 2-ethylhexyl terephthalate, bis 2-ethylhexyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-bis 2-ethylhexyl ester
Solubility Information Solubility in water: insoluble.
InChI Key RWPICVVBGZBXNA-UHFFFAOYSA-N
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)C(=O)OCC(CC)CCCC
IUPAC Name bis(2-ethylhexyl) benzene-1,4-dicarboxylate
Molecular Weight (g/mol) 390.55
PubChem CID 22932
Formula Weight 390.55
Percent Purity 97%
Chemical Name or Material Bis(2-ethylhexyl) terephthalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.