missing translation for 'onlineSavingsMsg'
Learn More

Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate, 95%

Product Code. 15490626
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490626 5 g 5g
15400636 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490626 Supplier Thermo Scientific Alfa Aesar Supplier No. H53378.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75203-51-9 | C26H24Cl6O8 | 677.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75203-51-9
Molecular Formula C26H24Cl6O8
Molecular Weight (g/mol) 677.17
MDL Number MFCD00012038
InChI Key PURKHUDOTFUVNG-UHFFFAOYSA-N
Synonym cppo, bis 2-carbopentyloxy-3,5,6-trichlorophenyl oxalate, bis 2,3,5-trichloro-6-pentyloxy carbonyl phenyl oxalate, bis 2-carbopentyloxy-3,5,6-trichloro-phenyl oxal, ethanedioic acid, bis 2,3,5-trichloro-6-pentyloxy carbonyl phenyl ester, pentyl 3,4,6-trichloro-2-2-oxo-2-2,3,5-trichloro-6-pentyloxy carbonyl phenoxy acetyl oxy benzoate, bis 2,3,5-trichloro-6-pentoxycarbonylphenyl oxalate, bis 2,3,5-trichloro-6-pentoxycarbonyl-phenyl oxalate
PubChem CID 3018575
IUPAC Name bis(2,3,5-trichloro-6-pentoxycarbonylphenyl) oxalate
SMILES CCCCCOC(=O)C1=C(C(=C(C=C1Cl)Cl)Cl)OC(=O)C(=O)OC2=C(C(=CC(=C2Cl)Cl)Cl)C(=O)OCCCCC

Specifications

Melting Point 76°C to 80°C
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 677.19
Percent Purity 95%
Chemical Name or Material Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.