missing translation for 'onlineSavingsMsg'
Learn More

Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine, 94%

Product Code. 11485620
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11485620 5 g 5g
11475620 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11485620

Brand: Thermo Scientific Alfa Aesar H25882.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 212713-08-1 | C30H46ClO2P | 505.12 g/mol

It is used as a reactant for asymmetric hydrogenation with iridium catalysts, chemo selective reaction with deprotonated 2-hydoxy-3,3,N-trimethylbutanamide. It is also used as a reactant for synthesis of phosphine- containing amino acids and peptide-based catalyst ligands, nonracemic phosphine-containing amino acids for palladium- catalyzed asymmetric allylic substitution reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reactant for asymmetric hydrogenation with iridium catalysts, chemo selective reaction with deprotonated 2-hydoxy-3,3,N-trimethylbutanamide. It is also used as a reactant for synthesis of phosphine- containing amino acids and peptide-based catalyst ligands, nonracemic phosphine-containing amino acids for palladium- catalyzed asymmetric allylic substitution reactions.

Solubility
Reacts with water.

Notes
Air & moisture sensitive. Materials to avoid is oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 212713-08-1
Molecular Formula C30H46ClO2P
Molecular Weight (g/mol) 505.12
MDL Number MFCD08458881
InChI Key VOGHMZHRQUWLRE-UHFFFAOYSA-N
Synonym bis 3,5-di-tert-butyl-4-methoxyphenyl chlorophosphine, acmc-20alnq, chlorobis 3,5-di-tert-butyl-4-methoxyphenyl phosphane, chlorobis 3,5-di-tert-butyl-4-methoxyphenyl phosphine, bis 3,5-di-tert-butyl-4-methoxyphenyl phosphinous chloride, bis 3,5-bis 1,1-dimethylethyl-4-methoxyphenyl phosphinous chloride, phosphinous chloride,bis 3,5-bis 1,1-dimethylethyl-4-methoxyphenyl-9ci
PubChem CID 53431072
IUPAC Name chloro-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane
SMILES CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC(=C(C(=C2)C(C)(C)C)OC)C(C)(C)C)Cl

Specifications

Quantity 5 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 505.13
Percent Purity 94%
Chemical Name or Material Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.