missing translation for 'onlineSavingsMsg'
Learn More

Bis(3-aminopropyl)phenylphosphine, Thermo Scientific Chemicals

Product Code. 11345928
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345928 10 g 10g
11355928 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345928 Supplier Thermo Scientific Alfa Aesar Supplier No. 010427.09
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6775-01-5 | Molecular Formula: C12H21N2P | Molecular Weight: 224.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6775-01-5
Molecular Formula C12H21N2P
Molecular Weight (g/mol) 224.29
MDL Number MFCD00014834
InChI Key OEHBMEXOPYWPKQ-UHFFFAOYSA-N
Synonym bis 3-aminopropyl phenylphosphine, 3,3'-phenylphosphinediyl bis propan-1-amine, 3-3-aminopropyl phenyl phosphanyl propan-1-amine, bis 3-aminopropyl phenyl phosphane, 1-propanamine,3,3'-phenylphosphinidene bis-9ci, bis 3-aminopropyl phenylphosphine, packaged under argon in resealable chemseal bottles
PubChem CID 138827
IUPAC Name 3-[3-aminopropyl(phenyl)phosphanyl]propan-1-amine
SMILES NCCCP(CCCN)C1=CC=CC=C1

Specifications

Density 1.03 g/mL
Boiling Point 144°C (1 mmHg)
Refractive Index 1.5728
Quantity 10 g
UN Number UN3267
Sensitivity Air Sensitive
Formula Weight 224.29
Physical Form Liquid
Chemical Name or Material Bis(3-aminopropyl)phenylphosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.