missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-aminophenyl) ether, 98%

Product Code. 11354579
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354579 50 g 50g
11412848 100 g 100g
11422848 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354579 Supplier Thermo Scientific Alfa Aesar Supplier No. A16815.18
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 101-80-4 | C12H12N2O | 200.241 g/mol

Bis(4-aminophenyl) ether is a monomer used as a cross-linking agent. It is also used in the preparation of poly(imideurea)s, polyimide silica composite membrane, which finds application in gas separation techniques. It is utilized in products such as wire enamels, coatings, adhesives, insulating varnishes, coated fabrics, flame-retardant fibers, oil sealants and retainers. Further, it is used in the preparation of polymer films for electrolyte membranes. In addition to this, it is used as an insulation for cables and printed circuits, and laminates and composite for aerospace vehicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-aminophenyl) ether is a monomer used as a cross-linking agent. It is also used in the preparation of poly(imideurea)s, polyimide silica composite membrane, which finds application in gas separation techniques. It is utilized in products such as wire enamels, coatings, adhesives, insulating varnishes, coated fabrics, flame-retardant fibers, oil sealants and retainers. Further, it is used in the preparation of polymer films for electrolyte membranes. In addition to this, it is used as an insulation for cables and printed circuits, and laminates and composite for aerospace vehicles.

Solubility
Soluble in acetone. Insoluble in water, benzene, carbon tetrachloride, ethanol and hot methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-80-4
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD00007863
InChI Key HLBLWEWZXPIGSM-UHFFFAOYSA-N
Synonym 4,4'-oxydianiline, 4,4'-diaminodiphenyl ether, 4,4-oxydianiline, 4-aminophenyl ether, 4,4'-oxybisbenzenamine, oxydianiline, 4-4-aminophenoxy aniline, diaminodiphenyl ether, benzenamine, 4,4'-oxybis, p,p'-oxydianiline
PubChem CID 7579
ChEBI CHEBI:34384
IUPAC Name 4-(4-aminophenoxy)aniline
SMILES C1=CC(=CC=C1N)OC2=CC=C(C=C2)N

Specifications

Melting Point 189°C to 192°C
Boiling Point 190°C (0.1 mmHg)
Flash Point 218°C (424°F)
Quantity 50 g
UN Number UN3077
Beilstein 475735
Solubility Information Soluble in acetone. Insoluble in water,benzene,carbon tetrachloride,ethanol and hot methanol.
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material Bis(4-aminophenyl) ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.