missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-chlorophenyl) sulfone, 99%

Product Code. 11403188
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403188 100 g 100g
11413188 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403188 Supplier Thermo Scientific Alfa Aesar Supplier No. A18938.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 80-07-9 | C12H8Cl2O2S | 287.15 g/mol

Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4'-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4′-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-07-9
Molecular Formula C12H8Cl2O2S
Molecular Weight (g/mol) 287.15
MDL Number MFCD00000619
InChI Key GPAPPPVRLPGFEQ-UHFFFAOYSA-N
Synonym 4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone
PubChem CID 6625
SMILES ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 147°C to 150°C
Boiling Point 249°C to 251°C (10 mmHg)
Flash Point 233°C (451°F)
Quantity 100 g
Beilstein 2052955
Solubility Information Insoluble in water.
IUPAC Name 1-chloro-4-(4-chlorophenyl)sulfonylbenzene
Formula Weight 287.17
Percent Purity 99%
Chemical Name or Material Bis(4-chlorophenyl) sulfone
compliance-icons compliance-icons
Product Identifier
  • Bis(4-chlorophenyl) sulphone
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Bis(4-chlorophenyl)sulphone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.