missing translation for 'onlineSavingsMsg'
Learn More

Bis-(4-hydroxy-1-naphthyl)phenylmethanol, Thermo Scientific™

Product Code. 10003460 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 10003460

Brand: Thermo Scientific Acros 301640250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6948-88-5
Molecular Formula C27H20O3
Molecular Weight (g/mol) 392.454
MDL Number MFCD00003979
InChI Key OUYLDJXFDLBCTQ-UHFFFAOYSA-N
Synonym bis 4-hydroxy-1-naphthyl phenylmethanol, 4,4'-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy-alpha-4-hydroxynaphthyl-alpha-phenylnaphthalene-1-methanol, 4-hydroxy-4-hydroxynaphthalen-1-yl-phenyl-methyl naphthalen-1-ol, bis 4-hydroxynaphthyl phenylmethan-1-ol, 4,4'-hydroxy phenyl methylene di 1-naphthol, 4,4-alpha-hydroxybenzylidene di-1-naphthol, bis-4-hydroxy-1 naphthyl-phenyl-methanol, 4,4-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy 4-hydroxynaphthalen-1-yl benzyl naphthalen-1-ol
PubChem CID 81382
IUPAC Name 4-[hydroxy-(4-hydroxynaphthalen-1-yl)-phenylmethyl]naphthalen-1-ol
SMILES C1=CC=C(C=C1)C(C2=CC=C(C3=CC=CC=C32)O)(C4=CC=C(C5=CC=CC=C54)O)O

Specifications

Melting Point 121.0°C to 125.0°C
Color Red to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Beilstein 06, IV, 7666
Formula Weight 392.46
Physical Form Powder
Chemical Name or Material Bis-(4-hydroxy-1-naphthyl)phenylmethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.