missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-hydroxy)benzophenone, TRC

Product Code. 30136902
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136902 5 g 5g
30010935 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136902 Supplier Toronto Research Chemicals Supplier No. TRCB4438005G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-99-4
Molecular Formula C13 H10 O3
Molecular Weight (g/mol) 214.22
Synonym 4,4'-Dihydroxybenzophenone, Benzophenone, 4,4'-dihydroxy- (7CI,8CI), Bis(4-hydroxyphenyl)methanone, 4,4'-Dihydroxybenzophenone, 4,4'-Dihydroxydiphenyl ketone, 4,4'-Hydroxybenzophenone, 4-DHB, Bis(4-hydroxyphenyl) ketone, Bis(p-hydroxy)benzophenone, HBP, HBP (ketone), NSC 2831, p,p'-Dihydroxybenzophenone
IUPAC Name bis(4-hydroxyphenyl)methanone
SMILES Oc1ccc(cc1)C(=O)c2ccc(O)cc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H10O3/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8,14-15H
Formula Weight 214.063
Chemical Name or Material 4,4'-Dihydroxybenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.