missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)zirconium hydridochloride, 95%, Thermo Scientific Chemicals

Product Code. 11446840
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446840 25 g 25g
11426840 1 g 1g
11436840 5 g 5g
11398176 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11446840 Supplier Thermo Scientific Alfa Aesar Supplier No. L15164.14
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Bis(cyclopentadienyl)zirconium hydridochloride is used in the conversion of amides to aldehydes. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(cyclopentadienyl)zirconium hydridochloride is used in the conversion of amides to aldehydes. It is also used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. It is sensitive to moisture, light and air. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37342-97-5
Molecular Formula C10H11ClZr-2
Molecular Weight (g/mol) 257.872
MDL Number MFCD02089401
InChI Key VPQMECJLNPUNGN-UHFFFAOYSA-M
Synonym zirconocene chloride hydride
PubChem CID 131718033
IUPAC Name cyclopenta-1,3-diene;hydride;zirconium(2+);chloride
SMILES [H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2]

Specifications

Melting Point >300°C
Quantity 25 g
UN Number UN3131
Sensitivity Air, light, and moisture sensitive
Merck Index 14,8396
Solubility Information Reacts with water.
Formula Weight 257.87
Percent Purity 95%
Chemical Name or Material Bis(cyclopentadienyl)zirconium hydridochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.