missing translation for 'onlineSavingsMsg'
Learn More

Bis(p-toluenesulfonyl)methane, 97%

Product Code. 11392479
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392479 1 g 1g
15433507 250 mg 250mg
11302489 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392479 Supplier Thermo Scientific Alfa Aesar Supplier No. H26617.03
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15310-28-8 | C15H16O4S2 | 324.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15310-28-8
Molecular Formula C15H16O4S2
Molecular Weight (g/mol) 324.41
MDL Number MFCD00032255
InChI Key XLOXYWLRAKCDQL-UHFFFAOYSA-N
Synonym ditosylmethane, bis-toluene-4-sulfonyl-methane, 1-methyl-4-4-methylbenzenesulfonyl methanesulfonyl benzene, 1,1'-methanediyldisulfonyl bis 4-methylbenzene, acmc-20allu, bis p-tolylsulfonyl methane, bis p-toluenesulfonyl methane, bis-toluene-4-sulfonyl methane, 1,1'-methylenedisulfonyl bis 4-methylbenzene, benzene,1,1'-methylenebis sulfonyl bis 4-methyl
PubChem CID 275690
IUPAC Name 1-methyl-4-[(4-methylphenyl)sulfonylmethylsulfonyl]benzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)CS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 137°C to 139°C
Quantity 1 g
Formula Weight 324.42
Percent Purity 97%
Chemical Name or Material Bis(p-toluenesulfonyl)methane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.