missing translation for 'onlineSavingsMsg'
Learn More

Bis(phenylsulfonyl)sulfide, 96%

Product Code. 10752301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10752301 1 g Glass bottle 1g
11945741 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752301 Supplier Thermo Scientific Acros Supplier No. 300830010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4388-22-1 | C12H10O4S3 | 314.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4388-22-1
Molecular Formula C12H10O4S3
Molecular Weight (g/mol) 314.39
MDL Number MFCD00094733
InChI Key YQUSJUJNDKUWAM-UHFFFAOYSA-N
Synonym bis phenylsulfonyl sulfide, benzenesulfonyl sulfide, benzenesulfonic thioanhydride, benzenesulfonothioic acid, anhydrosulfide, benzenesulfonic acid, thio-, anhydrosulfide, bis phenylsulfonyl thio, benzenesulfonyl sulfanylsulfonyl benzene, phso2 2s, benzenesulfonylthio sulfonylbenzene
PubChem CID 257516
IUPAC Name benzenesulfonylsulfanylsulfonylbenzene
SMILES O=S(=O)(SS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 129.0°C to 134.0°C
Color Pink to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 11,III,163
Formula Weight 314.41
Percent Purity 96%
Physical Form Crystals or Powder
Chemical Name or Material Bis(phenylsulfonyl)sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.