missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)acetylene, 99%

Product Code. 11463974
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463974 250 g 250g
11443974 10 g 10g
11453974 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463974 Supplier Thermo Scientific Alfa Aesar Supplier No. A11960.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14630-40-1 | C8H18Si2 | 170.402 g/mol

BTMSA is used as starting reagent in the synthesis of functionalized 4-R-1,2-bis(trimethylsilyl)benzenes. It is also used in the synthesis of (+)-brasilenyne2 and (β-diketanato)Ag(BTMSA). Bis(trimethylsilyl)acetylene is a ligand that binds to a central metal ion (e.g. Titanium) to create a coordination complex. One of its complexes, Permethyltitanocene-bis(trimethylsilyl)acetylene, is a catalyst for the head-to-tail dimerization of 1-alkynes (e.g. 3-Penten-1-yne [P227430]).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
BTMSA is used as starting reagent in the synthesis of functionalized 4-R-1,2-bis(trimethylsilyl)benzenes. It is also used in the synthesis of (+)-brasilenyne2 and (β-diketanato)Ag(BTMSA). Bis(trimethylsilyl)acetylene is a ligand that binds to a central metal ion (e.g. Titanium) to create a coordination complex. One of its complexes, Permethyltitanocene-bis(trimethylsilyl)acetylene, is a catalyst for the head-to-tail dimerization of 1-alkynes (e.g. 3-Penten-1-yne [P227430]).

Solubility
Not miscible or difficult to mix in water. Soluble in Chloroform and Methanol.

Notes
Store away from oxidizing agents. Do not store together with acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14630-40-1
Molecular Formula C8H18Si2
Molecular Weight (g/mol) 170.402
MDL Number MFCD00008276
InChI Key ZDWYFWIBTZJGOR-UHFFFAOYSA-N
Synonym bis trimethylsilyl acetylene, silane, 1,2-ethynediylbis trimethyl, 1,2-bis trimethylsilyl ethyne, btmsa, 1,2-bis-trimethylsilanyl-ethyne, ethyne-1,2-diylbis trimethylsilane, 2,5-disilahex-3-yne, 2,2,5,5-tetramethyl, trimethyl 2-trimethylsilyl ethynyl silane, silane, 1,1'-1,2-ethynediyl bis 1,1,1-trimethyl
PubChem CID 84564
IUPAC Name trimethyl(2-trimethylsilylethynyl)silane
SMILES C[Si](C)(C)C#C[Si](C)(C)C

Specifications

Melting Point 20°C to 24°C
Density 0.77
Boiling Point 134°C to 136°C
Flash Point 2°C (35°F)
Linear Formula (CH3)3SiC≡CSi(CH3)3
Refractive Index 1.427
Quantity 250 g
UN Number UN1993
Beilstein 906870
Solubility Information Not miscible or difficult to mix in water. Soluble in Chloroform and Methanol.
Formula Weight 170.4
Percent Purity 99%
Chemical Name or Material Bis (trimethylsilyl)acetylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.