missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)sulfide, tech.

Product Code. p-7059540
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11490111

Brand: Thermo Scientific Alfa Aesar 044818.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3385-94-2 | C6H18SSi2 | 178.44 g/mol

Bis(trimethylsilyl)sulfide is used to convert oxides and chlorides into the corresponding sulfides. It has been used in the conversion of aldehydes and ketones to the corresponding thiones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3385-94-2
Molecular Formula C6H18SSi2
Molecular Weight (g/mol) 178.44
MDL Number MFCD00014851
InChI Key RLECCBFNWDXKPK-UHFFFAOYSA-N
Synonym bis trimethylsilyl sulfide, hexamethyldisilthiane, hexamethyldisilathiane, disilathiane, hexamethyl, 1,1,1,3,3,3-hexamethyldisilathiane, disilthiane, hexamethyl, disilathiane, 1,1,1,3,3,3-hexamethyl, thiobis trimethylsilane, acmc-1ckj8
PubChem CID 76920
IUPAC Name trimethyl(trimethylsilylsulfanyl)silane
SMILES C[Si](C)(C)S[Si](C)(C)C

Specifications

Color Colorless
Density 0.851 g/mL
Boiling Point 162°C to 166°C
Flash Point 26°C (79°F)
Odor Pungent
Packaging Ampule
Linear Formula [(CH3)3Si]2S
Refractive Index 1.4598
Quantity 1 g
UN Number UN1993
Beilstein 1740046
Sensitivity Air Sensitive
Solubility Information Soluble in tetrahydrofuran and toluene. Hydrolyzes in water.
Formula Weight 178.44
Grade Technical
Physical Form Liquid
Chemical Name or Material Bis (trimethylsilyl)sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.