missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)nickel(II)chloride, 98%

Product Code. 10296700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10296700 10 g Glass bottle 10g
10723221 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10296700 Supplier Thermo Scientific Acros Supplier No. 217500100
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14264-16-5 | C36H30Cl2NiP2 | 654.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14264-16-5
Molecular Formula C36H30Cl2NiP2
Molecular Weight (g/mol) 654.18
MDL Number MFCD00009592 MFCD00003043 MFCD20489348
InChI Key VHQPRJDILRYZBG-UHFFFAOYSA-L
Synonym bis triphenylphosphine dichloronickel ii
PubChem CID 131664342
IUPAC Name nickel(2+);triphenylphosphane;dihydrochloride
SMILES Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >300.0°C
Color Gray to Green
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula [(C6H5)3P]2NiCl2
Quantity 10 g
Fieser 02,110; 09,147; 11,165; 16,118
Solubility Information Solubility in water: insoluble
Formula Weight 654.2
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Bis(triphenylphosphine)nickel(II)chloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.