missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)palladium(II) chloride, 98%

Product Code. 10333432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
2.5g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10333432 5 g Glass bottle 5g
10492061 2.5 g Glass bottle 2.5g
10649263 25 g Glass bottle 25g
11944361 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10333432 Supplier Thermo Scientific Acros Supplier No. undefined
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13965-03-2 | C36H30Cl2P2Pd | 701.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13965-03-2
Molecular Formula C36H30Cl2P2Pd
Molecular Weight (g/mol) 701.90
MDL Number MFCD00009593
InChI Key YNHIGQDRGKUECZ-UHFFFAOYSA-L
Synonym bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g
PubChem CID 131664180
IUPAC Name ethane;methane;palladium(2+);triphenylphosphane;dichloride
SMILES [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 260°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 14.8 to 15.5% (Pd)
Packaging Glass bottle
Linear Formula [(C6H5)3P]2PdCl2
Quantity 5 g
Fieser 06,59; 07,95; 08,151; 17,111
Solubility Information Solubility in water: insoluble
Formula Weight 701.89
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bis(triphenylphosphine)palladium(II) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Bis(triphenylphosphine)palladium(II) chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.