missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)palladium(II) chloride, for analysis, 15% Pd

Product Code. 11904361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11904361 25 g Glass bottle 25g
10113930 1 g Glass bottle 1g
10123930 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11904361 Supplier Thermo Scientific Acros Supplier No. 197320250
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13965-03-2 | C36H30Cl2P2Pd | 701.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13965-03-2
Molecular Formula C36H30Cl2P2Pd
Molecular Weight (g/mol) 701.90
MDL Number MFCD00009593
InChI Key YNHIGQDRGKUECZ-UHFFFAOYSA-L
Synonym bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g
PubChem CID 131664180
IUPAC Name ethane;methane;palladium(2+);triphenylphosphane;dichloride
SMILES [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 260°C
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula [(C6H5)3P]2PdCl2
Quantity 25 g
Fieser 06,59; 07,95; 08,151; 17,111
Solubility Information Solubility in water: insoluble.
Formula Weight 701.89
Percent Purity 15% Pd
Grade Analysis
Physical Form Powder
Chemical Name or Material Bis(triphenylphosphine)palladium(II) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Bis(triphenylphosphine)palladium(II) chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.