missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BIS-TRIS, 0.2M buffer soln., pH 6.5

Product Code. 15402959
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
This item is not returnable. View return policy

Product Code. 15402959

Brand: Thermo Scientific Chemicals J60127.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Color Colorless
Concentration 0.2 M
pH 6.5
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Chemical Name or Material BIS-TRIS
Hazard Statement
  • EUH210-Safety data sheet available on request.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.