missing translation for 'onlineSavingsMsg'
Learn More

Melford bis-TRIS

Product Code. 30018127
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018127 500 g 500g
30018125 100 g 100g
30018126 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018127 Supplier Melford Supplier No. B75000500.0
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Substance Name: bis-(2-hydroxyethyl)aminotris(hydroxymethyl)methane, CAS Number: 6976-37-0, Molecular Formula: C8H19NO5, Molecular Weight:?209, Chemical Purity:?>99%, Soluble in: Water, Store at:?Room Temperature

Specifications

CAS 6976-37-0
Quantity 500 g
Molecular Formula C8H19NO5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.