missing translation for 'onlineSavingsMsg'
Learn More

Bis-Tris, ≥98% (Dry Basis), Ultrapure

Product Code. 15815098 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15815098 100 g Plastic bottle 100g
15825098 500 g Plastic bottle 500g
15805268 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15815098 Supplier Thermo Scientific Supplier No. J12112.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym bis[2 to Hydroxyethyl]imino to tris[hydroxymethyl]methane
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Melting Point 102°C to 105°C
Physical Form Crystalline Powder
Quantity 100 g
Linear Formula (HOCH2CH2)2NC(CH2OH)3
Packaging Plastic bottle
Beilstein 2205275
Formula Weight 209.24
Percent Purity ≥98%
Assay Dry basis
Chemical Name or Material bis[2-Hydroxyethyl]imino-tris[hydroxymethyl]methane
compliance-icons
Product Identifier
  • Bis-Tris, Ultrapure, Thermo Scientific
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Bis-Tris

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.