missing translation for 'onlineSavingsMsg'
Learn More

Bis-Tris, ≥98% (Dry Basis), Ultrapure

Product Code. 15825098 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 15825098

Brand: Thermo Scientific J12112.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym bis[2 to Hydroxyethyl]imino to tris[hydroxymethyl]methane
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Melting Point 102°C to 105°C
Physical Form Crystalline Powder
Quantity 500 g
Linear Formula (HOCH2CH2)2NC(CH2OH)3
Packaging Plastic bottle
Beilstein 2205275
Formula Weight 209.24
Percent Purity ≥98%
Chemical Name or Material bis[2-Hydroxyethyl]imino-tris[hydroxymethyl]methane
compliance-icons
Product Identifier
  • Bis-Tris, Ultrapure, Thermo Scientific
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Bis-Tris

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.