missing translation for 'onlineSavingsMsg'
Learn More

bis-Tris, ≥99%, MP Biomedicals™

Product Code. 11358609 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358609 1 kg 1kg
11328609 25 g 25g
11338609 100 g 100g
11348609 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11358609 Supplier MP Biomedicals Supplier No. 0210103891
  • ONLINE EXCLUSIVE PRICE
    1360.00 €
    1286.25 € / 1kg
    Save 73.75 €
    5% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Exhibits a very small shift in dissociation constant with respect to temperature

  • Zwitterionic buffer
  • Buffering agent
  • White Powder

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 kg
Solution Type Buffer
Assay Percent Range >99%
Synonym 2,2-bis(Hydroxymethyl)-2,2',2”-nitrilotriethanol, 2, 2-bis(Hydroxymethyl)-2, 2'
Formula Weight 209.2
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material bis-Tris
compliance-icons
Product Identifier
  • Bis-Tris
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.