missing translation for 'onlineSavingsMsg'
Learn More

Bis-Tris, Fisher BioReagents

Product Code. 10041463
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10041463 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10041463 Supplier Fisher BioReagents Supplier No. BP301100
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Commonly used buffering agent

This biological buffer has a usable pH range of 5.8 to 7.2.
TRUSTED_SUSTAINABILITY

Specifications

CAS 6976-37-0
Melting Point 103°C
Color White
pH 8.8 to 9.6
Physical Form Powder or Solid
Quantity 100 g
Assay Percent Range ≥99 %
Identification Pass Test
Molecular Formula C8H19NO5
Linear Formula (HOCH2CH2)2NC(CH2OH)3
Packaging Poly Bottle
Vapor Pressure Negligible
Synonym 2,2-Bis(hydroxymethyl)-2,2?,2?-nitrilotriethanol, 2, 2-Bis(hydroxymethyl)-2, 2?, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
SMILES C(CO)N(CCO)C(CO)(CO)CO
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 209.242
PubChem CID 81462
ChEBI CHEBI:41250
Formula Weight 209.24
Percent Purity ≥99%
Chemical Name or Material Bis-Tris
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.