missing translation for 'onlineSavingsMsg'
Learn More

Bisacodyl, MedChemExpress

Product Code. 16450998
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16450998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16450998 Supplier Medchem Express Supplier No. HYB0557
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bisacodyl is a stimulant laxative agent that works directly on the colon to produce a bowel movement. Bisacodyl increases the secretion of PGE2 by direct activation of colon macrophages. PGE2 acts as a paracrine factor and decreases the expression of AQP3 in the colon, which inhibits water transfer from the luminal to the vascular side and leads to a laxative effect.

Bisacodyl is a stimulant laxative agent that works directly on the colon to produce a bowel movement. Bisacodyl increases the secretion of PGE2 by direct activation of colon macrophages. PGE2 acts as a paracrine factor and decreases the expression of AQP3 in the colon, which inhibits water transfer from the luminal to the vascular side and leads to a laxative effect.

Specifications

Chemical Name or Material Bisacodyl
CAS 603-50-9
Color White
Purity Grade Notes Research
Molecular Formula C22H19NO4
Quantity 5 g
Solubility Information DMSO : 50 mg/mL (138.35 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.28 mM; Need ultrasonic)
SMILES CC(OC1=CC=C(C(C2=CC=C(OC(C)=O)C=C2)C3=NC=CC=C3)C=C1)=O
Molecular Weight (g/mol) 361.39
Formula Weight 361.39
Percent Purity 99.18%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.